Assay Details
| Gene Symbol: Nrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872081.2 | XP_004872138.1 | 8 - 9 | 764 | 81 | |
| XM_021250092.1 | XP_021105751.1 | 9 - 10 | 948 | 81 | ||
| XM_021250093.1 | XP_021105752.1 | 8 - 9 | 761 | 81 | ||
| XM_021250094.1 | XP_021105753.1 | 9 - 10 | 852 | 81 | ||
| XM_021250095.1 | XP_021105754.1 | 7 - 8 | 672 | 81 | ||
| XM_021250096.1 | XP_021105755.1 | 9 - 10 | 948 | 81 | ||
| XM_021250097.1 | XP_021105756.1 | 9 - 10 | 948 | 81 | ||
More Information
Set Membership: |
|

Nrg4
Genomic Map