Assay Details
| Gene Symbol: LOC106008787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013069594.2 | XP_012925048.2 | 2 - 3 | 198 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC106008787| Gene Symbol: LOC106008787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013069594.2 | XP_012925048.2 | 2 - 3 | 198 | 78 | |
Set Membership: |
|