Assay Details
| Gene Symbol: Pla2g10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013076316.2 | XP_012931770.1 | 1 - 2 | 197 | 93 | |
| XM_013076318.2 | XP_012931772.1 | 1 - 2 | 197 | 93 | ||
More Information
Set Membership: |
|

Pla2g10
Genomic Map