Assay Details
| Gene Symbol: LOC101726216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021254558.1 | XP_021110217.1 | 14 - 15 | 1245 | 94 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101726216| Gene Symbol: LOC101726216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021254558.1 | XP_021110217.1 | 14 - 15 | 1245 | 94 | |