Assay Details
| Gene Symbol: LOC110350810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021265106.1 | XP_021120765.1 | 2 - 3 | 249 | 50 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC110350810| Gene Symbol: LOC110350810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021265106.1 | XP_021120765.1 | 2 - 3 | 249 | 50 | |
Set Membership: |
|