Assay Details
| Gene Symbol: Kif3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840580.3 | XP_004840637.1 | 7 - 8 | 2145 | 70 | |
| XM_013070658.2 | XP_012926112.1 | 8 - 9 | 2174 | 70 | ||
| XM_021254888.1 | XP_021110547.1 | 7 - 8 | 2185 | 70 | ||
| XM_021254889.1 | XP_021110548.1 | 7 - 8 | 2193 | 70 | ||
More Information
Set Membership: |
|

Kif3b
Genomic Map