Assay Details
| Gene Symbol: LOC101704762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841162.2 | XP_004841219.1 | 1 - 2 | 280 | 73 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101704762| Gene Symbol: LOC101704762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841162.2 | XP_004841219.1 | 1 - 2 | 280 | 73 | |