Assay Details
| Gene Symbol: Cetp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843186.2 | XP_004843243.1 | 7 - 8 | 676 | 97 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cetp| Gene Symbol: Cetp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843186.2 | XP_004843243.1 | 7 - 8 | 676 | 97 | |