Assay Details
| Gene Symbol: Cap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847637.3 | XP_004847694.1 | 7 - 8 | 794 | 58 | |
| XM_021260597.1 | XP_021116256.1 | 6 - 7 | 533 | 58 | ||
More Information
Set Membership: |
|

Cap2
Genomic Map