Assay Details
| Gene Symbol: Yrdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850997.3 | XP_004851054.1 | 4 - 5 | 790 | 60 | |
| XM_004850998.3 | XP_004851055.1 | 3 - 4 | 670 | 60 | ||
More Information
Set Membership: |
|

Yrdc
Genomic Map