Assay Details
| Gene Symbol: LOC101707238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853654.3 | XP_004853711.1 | 6 - 7 | 593 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101707238| Gene Symbol: LOC101707238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853654.3 | XP_004853711.1 | 6 - 7 | 593 | 95 | |
Set Membership: |
|