Assay Details
| Gene Symbol: Capza1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853732.3 | XP_004853789.1 | 5 - 6 | 458 | 93 | |
| XM_004853733.3 | XP_004853790.1 | 5 - 6 | 458 | 93 | ||
| XM_004853734.3 | XP_004853791.1 | 4 - 5 | 350 | 93 | ||
More Information
Set Membership: |
|

Capza1
Genomic Map