Assay Details
| Gene Symbol: Smim19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855274.3 | XP_004855331.1 | 2 - 3 | 224 | 99 | |
| XM_004855275.3 | XP_004855332.1 | 2 - 3 | 271 | 99 | ||
More Information
Set Membership: |
|

Smim19
Genomic Map