Assay Details
| Gene Symbol: Helb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861096.3 | XP_004861153.1 | 6 - 7 | 1954 | 68 | |
| XM_021241553.1 | XP_021097212.1 | 6 - 7 | 1840 | 68 | ||
| XM_021241554.1 | XP_021097213.1 | 5 - 6 | 1465 | 68 | ||
| XM_021241555.1 | XP_021097214.1 | 6 - 7 | 1954 | 68 | ||
More Information
Set Membership: |
|

Helb
Genomic Map