Assay Details
| Gene Symbol: Kcnh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861998.3 | XP_004862055.1 | 7 - 8 | 1484 | 63 | |
| XM_004861999.3 | XP_004862056.1 | 7 - 8 | 1403 | 63 | ||
| XM_021242471.1 | XP_021098130.1 | 7 - 8 | 1800 | 63 | ||
| XM_021242472.1 | XP_021098131.1 | 7 - 8 | 1719 | 63 | ||
| XM_021242473.1 | XP_021098132.1 | 7 - 8 | 1726 | 63 | ||
| XM_021242474.1 | XP_021098133.1 | 7 - 8 | 1652 | 63 | ||
| XM_021242475.1 | XP_021098134.1 | 4 - 5 | 1132 | 63 | ||
| XM_021242476.1 | XP_021098135.1 | 7 - 8 | 1800 | 63 | ||
More Information
Set Membership: |
|

Kcnh1
Genomic Map