Assay Details
| Gene Symbol: Nr4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862916.3 | XP_004862973.2 | 3 - 4 | 1206 | 57 | |
| XM_004862917.3 | XP_004862974.1 | 3 - 4 | 1203 | 57 | ||
| XM_021243064.1 | XP_021098723.1 | 3 - 4 | 1104 | 57 | ||
More Information
Set Membership: |
|

Nr4a1
Genomic Map