Assay Details
| Gene Symbol: Gpr157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863924.2 | XP_004863981.1 | 2 - 3 | 698 | 64 | |
| XM_021243553.1 | XP_021099212.1 | 2 - 3 | 698 | 64 | ||
More Information
Set Membership: |
|

Gpr157
Genomic Map