Assay Details
| Gene Symbol: Crlf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867305.3 | XP_004867362.2 | 3 - 4 | 740 | 104 | |
| XM_004867306.3 | XP_004867363.2 | 3 - 4 | 732 | 104 | ||
More Information
Set Membership: |
|

Crlf2
Genomic Map