Assay Details
| Gene Symbol: Ddo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873982.3 | XP_004874039.1 | 1 - 2 | 1127 | 73 | |
| XM_013068961.2 | XP_012924415.1 | 1 - 2 | 985 | 73 | ||
| XM_021251532.1 | XP_021107191.1 | 1 - 2 | 203 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004873982.3, XM_013068961.2, XM_021251532.1 transcripts.
Set Membership: |
|

Ddo
Genomic Map