Assay Details
| Gene Symbol: Ddo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873982.3 | XP_004874039.1 | 4 - 5 | 1568 | 87 | |
| XM_013068960.1 | XP_012924414.1 | 3 - 4 | 465 | 87 | ||
| XM_013068961.2 | XP_012924415.1 | 4 - 5 | 1480 | 87 | ||
| XM_013068962.2 | XP_012924416.1 | 4 - 5 | 672 | 87 | ||
| XM_021251529.1 | XP_021107188.1 | 4 - 5 | 757 | 87 | ||
| XM_021251530.1 | XP_021107189.1 | 3 - 4 | 475 | 87 | ||
| XM_021251531.1 | XP_021107190.1 | 3 - 4 | 448 | 87 | ||
More Information
Set Membership: |
|

Ddo
Genomic Map