Assay Details
| Gene Symbol: Timm23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874516.3 | XP_004874573.1 | 1 - 2 | 228 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Timm23b| Gene Symbol: Timm23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874516.3 | XP_004874573.1 | 1 - 2 | 228 | 94 | |
Set Membership: |
|