Assay Details
| Gene Symbol: Timm23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874516.3 | XP_004874573.1 | 3 - 4 | 384 | 63 | |
| XM_004874517.3 | XP_004874574.1 | 3 - 4 | 312 | 63 | ||
| XM_021251944.1 | XP_021107603.1 | 4 - 5 | 581 | 63 | ||
More Information
Set Membership: |
|

Timm23b
Genomic Map