Assay Details
| Gene Symbol: Rimbp3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004921074.2 | XP_004921131.1 | 1 | 4926 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rimbp3c| Gene Symbol: Rimbp3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004921074.2 | XP_004921131.1 | 1 | 4926 | 93 | |
Set Membership: |
|