Assay Details
| Gene Symbol: Prg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013067471.2 | XP_012922925.1 | 4 - 5 | 583 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prg3| Gene Symbol: Prg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013067471.2 | XP_012922925.1 | 4 - 5 | 583 | 67 | |
Set Membership: |
|