Assay Details
| Gene Symbol: Wbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013072060.2 | XP_012927514.1 | 4 - 5 | 568 | 76 | |
| XM_021257916.1 | XP_021113575.1 | 4 - 5 | 568 | 76 | ||
More Information
Set Membership: |
|

Wbp4
Genomic Map