Assay Details
| Gene Symbol: Majin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013075043.2 | XP_012930497.1 | 8 - 9 | 821 | 87 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Majin| Gene Symbol: Majin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013075043.2 | XP_012930497.1 | 8 - 9 | 821 | 87 | |