Assay Details
| Gene Symbol: LOC101706828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021243896.1 | XP_021099555.1 | 7 - 8 | 771 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101706828| Gene Symbol: LOC101706828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021243896.1 | XP_021099555.1 | 7 - 8 | 771 | 88 | |
Set Membership: |
|