Assay Details
| Gene Symbol: Crkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021252234.1 | XP_021107893.1 | 2 - 3 | 725 | 58 | |
| XM_021252235.1 | XP_021107894.1 | 1 - 2 | 825 | 58 | ||
More Information
Set Membership: |
|

Crkl
Genomic Map