Assay Details
Gene Symbol: Slc10a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021255981.1 | XP_021111640.1 | 1 | 2620 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
