Assay Details
| Gene Symbol: Tead3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021259946.1 | XP_021115605.1 | 11 - 12 | 1204 | 54 | |
| XM_021259947.1 | XP_021115606.1 | 10 - 11 | 1131 | 54 | ||
| XM_021259948.1 | XP_021115607.1 | 10 - 11 | 1137 | 54 | ||
| XM_021259949.1 | XP_021115608.1 | 11 - 12 | 1253 | 54 | ||
More Information
Set Membership: |
|

Tead3
Genomic Map