Assay Details
| Gene Symbol: Atp5s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835570.3 | XP_004835627.1 | 2 - 3 | 732 | 79 | |
| XM_004835572.3 | XP_004835629.2 | 2 - 3 | 276 | 79 | ||
| XM_021262530.1 | XP_021118189.1 | 2 - 3 | 344 | 79 | ||
| XM_021262533.1 | XP_021118192.1 | 2 - 3 | 302 | 79 | ||
More Information
Set Membership: |
|

Atp5s
Genomic Map