Assay Details
| Gene Symbol: Atp5s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835570.3 | XP_004835627.1 | 3 - 4 | 935 | 114 | |
| XM_004835572.3 | XP_004835629.2 | 3 - 4 | 479 | 114 | ||
| XM_021262530.1 | XP_021118189.1 | 3 - 4 | 547 | 114 | ||
| XM_021262533.1 | XP_021118192.1 | 3 - 4 | 505 | 114 | ||
More Information
Set Membership: |
|

Atp5s
Genomic Map