Assay Details
| Gene Symbol: Abhd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840045.3 | XP_004840102.1 | 2 - 3 | 330 | 86 | |
| XM_004840046.3 | XP_004840103.1 | 2 - 3 | 330 | 86 | ||
| XM_013070503.2 | XP_012925957.1 | 2 - 3 | 330 | 86 | ||
| XM_013070504.2 | XP_012925958.1 | 2 - 3 | 330 | 86 | ||
| XM_021254399.1 | XP_021110058.1 | 2 - 3 | 330 | 86 | ||
More Information
Set Membership: |
|

Abhd11
Genomic Map