Assay Details
| Gene Symbol: Lcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842998.3 | XP_004843055.1 | 5 - 6 | 933 | 86 | |
| XM_021256829.1 | XP_021112488.1 | 4 - 5 | 746 | 86 | ||
More Information
Set Membership: |
|

Lcat
Genomic Map