Assay Details
| Gene Symbol: Sept3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845493.3 | XP_004845550.2 | 3 - 4 | 490 | 67 | |
| XM_004845494.3 | XP_004845551.2 | 3 - 4 | 490 | 67 | ||
| XM_004845495.3 | XP_004845552.1 | 3 - 4 | 490 | 67 | ||
| XM_004845496.3 | XP_004845553.1 | 4 - 5 | 625 | 67 | ||
| XM_013072556.2 | XP_012928010.1 | 3 - 4 | 690 | 67 | ||
| XM_021258906.1 | XP_021114565.1 | 3 - 4 | 490 | 67 | ||
| XM_021258907.1 | XP_021114566.1 | 3 - 4 | 490 | 67 | ||
| XM_021258908.1 | XP_021114567.1 | 4 - 5 | 561 | 67 | ||
| XM_021258909.1 | XP_021114568.1 | 3 - 4 | 476 | 67 | ||
More Information
Set Membership: |
|

Sept3
Genomic Map