Assay Details
| Gene Symbol: Eif3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845685.3 | XP_004845742.1 | 14 - 15 | 1865 | 98 | |
| XM_004845686.3 | XP_004845743.1 | 14 - 15 | 1860 | 98 | ||
| XM_021259049.1 | XP_021114708.1 | 13 - 14 | 1649 | 98 | ||
More Information
Set Membership: |
|

Eif3d
Genomic Map