Assay Details
| Gene Symbol: Mylk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847783.2 | XP_004847840.1 | 16 - 17 | 7431 | 75 | |
| XM_021260760.1 | XP_021116419.1 | 16 - 17 | 7429 | 75 | ||
More Information
Set Membership: |
|

Mylk4
Genomic Map