Assay Details
| Gene Symbol: LOC101711173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849828.2 | XP_004849885.1 | 9 - 10 | 1272 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101711173| Gene Symbol: LOC101711173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849828.2 | XP_004849885.1 | 9 - 10 | 1272 | 99 | |
Set Membership: |
|