Assay Details
| Gene Symbol: Eri2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855896.3 | XP_004855953.1 | 7 - 8 | 857 | 79 | |
| XM_021237283.1 | XP_021092942.1 | 7 - 8 | 854 | 79 | ||
| XM_021237284.1 | XP_021092943.1 | 7 - 8 | 854 | 79 | ||
| XM_021237285.1 | XP_021092944.1 | 7 - 8 | 854 | 79 | ||
More Information
Set Membership: |
|

Eri2
Genomic Map