Assay Details
| Gene Symbol: Dstyk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862117.3 | XP_004862174.1 | 12 - 13 | 2789 | 58 | |
| XM_021242232.1 | XP_021097891.1 | 12 - 13 | 2687 | 58 | ||
| XM_021242233.1 | XP_021097892.1 | 11 - 12 | 2612 | 58 | ||
| XM_021242234.1 | XP_021097893.1 | 12 - 13 | 2562 | 58 | ||
More Information
Set Membership: |
|

Dstyk
Genomic Map