Assay Details
| Gene Symbol: Ssc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866640.2 | XP_004866697.2 | 12 - 13 | 3216 | 58 | |
| XM_021245784.1 | XP_021101443.1 | 12 - 13 | 3213 | 58 | ||
| XM_021245785.1 | XP_021101444.1 | 12 - 13 | 3207 | 58 | ||
| XM_021245786.1 | XP_021101445.1 | 11 - 12 | 2901 | 58 | ||
| XM_021245787.1 | XP_021101446.1 | 11 - 12 | 2892 | 58 | ||
More Information
Set Membership: |
|

Ssc5d
Genomic Map