Assay Details
| Gene Symbol: Kcnc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866833.2 | XP_004866890.1 | 3 - 4 | 2935 | 96 | |
| XM_021246031.1 | XP_021101690.1 | 3 - 4 | 2935 | 96 | ||
| XM_021246032.1 | XP_021101691.1 | 3 - 4 | 2935 | 96 | ||
More Information
Set Membership: |
|

Kcnc3
Genomic Map