Assay Details
| Gene Symbol: Kcnc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866833.2 | XP_004866890.1 | 4 - 5 | 3062 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnc3| Gene Symbol: Kcnc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866833.2 | XP_004866890.1 | 4 - 5 | 3062 | 75 | |
Set Membership: |
|