Assay Details
| Gene Symbol: LOC101705383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868063.3 | XP_004868120.1 | 11 - 12 | 1990 | 75 | |
| XM_013066756.2 | XP_012922210.1 | 11 - 12 | 1926 | 75 | ||
| XM_021247070.1 | XP_021102729.1 | 11 - 12 | 2411 | 75 | ||
| XM_021247071.1 | XP_021102730.1 | 12 - 13 | 2049 | 75 | ||
| XM_021247072.1 | XP_021102731.1 | 11 - 12 | 2269 | 75 | ||
| XM_021247073.1 | XP_021102732.1 | 11 - 12 | 1991 | 75 | ||
More Information
Set Membership: |
|

LOC101705383
Genomic Map