Assay Details
| Gene Symbol: Gpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868429.2 | XP_004868486.1 | 7 - 8 | 1322 | 61 | |
| XM_004868430.3 | XP_004868487.1 | 7 - 8 | 2544 | 61 | ||
| XM_004868431.2 | XP_004868488.1 | 7 - 8 | 2428 | 61 | ||
| XM_004868432.2 | XP_004868489.1 | 7 - 8 | 1718 | 61 | ||
| XM_004868433.2 | XP_004868490.1 | 7 - 8 | 1239 | 61 | ||
More Information
Set Membership: |
|

Gpc1
Genomic Map