Assay Details
| Gene Symbol: Crlf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873382.2 | XP_004873439.1 | 2 - 3 | 418 | 72 | |
| XM_021251138.1 | XP_021106797.1 | 2 - 3 | 418 | 72 | ||
More Information
Set Membership: |
|

Crlf1
Genomic Map