Assay Details
| Gene Symbol: Zfand6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874812.2 | XP_004874869.1 | 7 - 8 | 1076 | 62 | |
| XM_004874813.3 | XP_004874870.1 | 7 - 8 | 982 | 62 | ||
| XM_004874816.2 | XP_004874873.1 | 6 - 7 | 913 | 62 | ||
| XM_004874817.3 | XP_004874874.1 | 6 - 7 | 790 | 62 | ||
| XM_004874820.2 | XP_004874877.1 | 5 - 6 | 677 | 62 | ||
| XM_004874821.3 | XP_004874878.1 | 5 - 6 | 582 | 62 | ||
| XM_013069276.2 | XP_012924730.1 | 8 - 9 | 1027 | 62 | ||
| XM_013069277.2 | XP_012924731.1 | 7 - 8 | 860 | 62 | ||
| XM_013069278.2 | XP_012924732.1 | 6 - 7 | 736 | 62 | ||
| XM_013069279.2 | XP_012924733.1 | 7 - 8 | 981 | 62 | ||
| XM_021252183.1 | XP_021107842.1 | 6 - 7 | 830 | 62 | ||
More Information
Set Membership: |
|

Zfand6
Genomic Map