Assay Details
| Gene Symbol: LOC101700519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013065553.2 | XP_012921007.1 | 3 - 4 | 857 | 59 | |
| XM_013065554.2 | XP_012921008.1 | 3 - 4 | 857 | 59 | ||
More Information
Set Membership: |
|

LOC101700519
Genomic Map