Assay Details
| Gene Symbol: LOC101703473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021246369.1 | XP_021102028.1 | 2 - 3 | 781 | 65 | |
| XM_021246370.1 | XP_021102029.1 | 2 - 3 | 791 | 65 | ||
| XM_021246371.1 | XP_021102030.1 | 2 - 3 | 791 | 65 | ||
| XM_021246372.1 | XP_021102031.1 | 2 - 3 | 791 | 65 | ||
More Information
Set Membership: |
|

LOC101703473
Genomic Map