Assay Details
| Gene Symbol: LOC101703473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021246369.1 | XP_021102028.1 | 3 - 4 | 1492 | 99 | |
| XM_021246372.1 | XP_021102031.1 | 3 - 4 | 1502 | 99 | ||
More Information
Set Membership: |
|

LOC101703473
Genomic Map